Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dsor1
Entrez Gene ID
31872
Gene Name
Downstream of raf1
Gene Aliases
CG15793, D-MEK, D-MEK/Dsor, D-Mek, D-SOR, D-Sor, D-mek, D-sor, D-sor-1, DMEK-1, DRODSOR1, DSOR, DSOR1, DSORT, DSor, DSor1, Dmek, Dmel\CG15793, Dmel_CG15793, Dsor, EK1-1, MAPKK, MEK, MEK/Dsor1, MEK1/2, Mek, Mek1/2, SOR, Sor, Su(Raf)34B, dMEK, dMek, dSor, dSor1, dsor1, mek, sor, sor/MEK1
Chromosome Location
Chr.X: 9247342 - 9250037 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010269
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298115.1 NP_001285044.1 3 - 4 1237 56
NM_078543.4 NP_511098.1 3 - 4 1533 56
GenBank mRNA AY058692.1 - 3 - 4 1080 56

View Full Product Details