Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- lz
- Entrez Gene ID
-
31883
- Gene Name
- lozenge
- Gene Aliases
-
CG1689, Dmel\CG1689, Dmel_CG1689, LZ, Lozenge, Lz, amx, fs(1)A1569, fs(1)M69, spe
- Chromosome Location
-
Chr.X: 9284649 - 9303636 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002576
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078544.3 |
NP_511099.2 |
5 - 6 |
1798 |
81 |
View Full Product Details