Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lilli
Entrez Gene ID
33496
Gene Name
lilliputian
Gene Aliases
154454_at, 189Y, AFF4, BEST:GH09955, BcDNA:GM02019, CG8817, Dmel\CG8817, Dmel_CG8817, Lilli, S(ls)4, SS2-1, SY2-1, Su(Raf)2A, Su(Raf)2a, dAFF, dAFF4, l(2)00632, l(2)23Cc, l(2)K00632, l(2)K07920, l(2)k07920, l(2)k16124
Chromosome Location
Chr.2L: 2885929 - 2954406 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041111
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144300.2 NP_001137772.1 3 - 4 1244 60
NM_001144301.1 NP_001137773.1 4 - 5 1584 60
NM_001144302.2 NP_001137774.1 3 - 4 1353 60
NM_001144303.1 NP_001137775.1 5 - 6 2888 60
NM_001273044.1 NP_001259973.1 4 - 5 3041 60
NM_001347818.1 NP_001334729.1 4 - 5 3041 60
NM_078740.3 NP_523464.1 5 - 6 3804 60
NM_164516.2 NP_722863.1 6 - 7 3972 60
NM_164517.1 NP_722864.1 5 - 6 2693 60
GenBank mRNA AF289034.1 - 6 - 7 3949 60
AF293971.1 - 5 - 6 3778 60

View Full Product Details