Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
qkr54B
Entrez Gene ID
36966
Gene Name
quaking related 54B
Gene Aliases
CG4816, Dmel\CG4816, Dmel_CG4816, QKR54B, Qkr54B, Qrk54B, SAM, SAM50, Sam50, p62GAP, qrk54B
Chromosome Location
Chr.2R: 17469624 - 17475129 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0022987
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274121.1 NP_001261050.1 2 - 3 401 63
NM_079048.4 NP_523772.1 2 - 3 766 63
NM_166230.3 NP_725679.1 2 - 3 411 63
NM_166231.3 NP_725680.2 2 - 3 401 63
GenBank mRNA AF096272.1 - 2 - 3 365 63
AY095201.1 - 2 - 3 393 63

View Full Product Details