Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mapmodulin
Entrez Gene ID
37043
Gene Name
CG5784-PA
Gene Aliases
CG5784, CT18148, CT42180, Dmel\CG5784, Dmel_CG5784, anon-EST:fe3A2
Chromosome Location
Chr.2R: 17866178 - 17872015 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034282
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079056.4 NP_523780.1 4 - 5 753 186
NM_166258.3 NP_725732.1 3 - 4 822 186
GenBank mRNA AY118324.1 - 4 - 5 745 186
BT012474.1 - 3 - 4 813 186

View Full Product Details