Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ena
- Entrez Gene ID
-
37201
- Gene Name
- enabled
- Gene Aliases
-
CG15112, Dmel\CG15112, Dmel_CG15112, ENA/VASP, Ena, Enb, VASP, Vasp, enb, l(2)02029
- Chromosome Location
-
Chr.2R: 19141632 - 19163705 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000578
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166331.3 |
NP_725859.2 |
1 - 2 |
695 |
54 |
View Full Product Details