Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fem-1
Entrez Gene ID
37344
Gene Name
CG9025-PA
Gene Aliases
CG9025, Dmel\CG9025, Dmel_CG9025, dFEM-1, dFEM1B
Chromosome Location
Chr.2R: 20662474 - 20667306 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034542
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137664.3 NP_611508.1 4 - 5 1831 66
NM_166408.2 NP_725993.1 5 - 6 1638 66
GenBank mRNA BT133040.1 - 4 - 5 1768 66

View Full Product Details