Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exu
Entrez Gene ID
37345
Gene Name
exuperantia
Gene Aliases
CG8994, Dmel\CG8994, Dmel_CG8994, Exu, bs30h09.y1, exu1
Chromosome Location
Chr.2R: 20667427 - 20671083 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000615
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169747.2 NP_001163218.1 3 - 4 447 95
NM_166410.3 NP_725995.1 3 - 4 769 95
NM_166411.3 NP_725996.1 2 - 3 437 95
NM_166412.3 NP_725997.1 2 - 3 437 95
GenBank mRNA AY051739.1 - 2 - 3 438 95
BT126187.1 - 3 - 4 461 95

View Full Product Details