Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vrp1
Entrez Gene ID
37521
Gene Name
Verprolin 1
Gene Aliases
CG13503, D-WIP, DWIP, DWip, Dm_Vrp1, Dmel\CG13503, Dmel_CG13503, Sltr, WIP, Wip, dWIP, sltr, solas, vrp, vrp1, wip
Chromosome Location
Chr.2R: 22106107 - 22122562 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0243516
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103936.2 NP_001097406.1 6 - 7 2055 62
NM_001169775.2 NP_001163246.1 6 - 7 1495 62
NM_001169776.2 NP_001163247.1 6 - 7 1577 62
NM_001169777.2 NP_001163248.1 6 - 7 2055 62
NM_001169778.2 NP_001163249.1 6 - 7 1577 62
NM_137796.3 NP_611640.3 6 - 7 1495 62
NM_166507.2 NP_726159.1 6 - 7 1768 62
NM_166508.2 NP_726160.1 6 - 7 2055 62
NM_166509.2 NP_726161.1 7 - 8 1918 62
NM_166510.2 NP_726162.1 6 - 7 1577 62
NM_166511.2 NP_726163.1 6 - 7 1563 62
GenBank mRNA AY094731.1 - 6 - 7 2069 62

View Full Product Details