Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tektin-A
Entrez Gene ID
34934
Gene Name
Tektin A
Gene Aliases
BG:DS02252.2, CG4767, Dmel\CG4767, Dmel_CG4767, anon-WO0140519.235
Chromosome Location
Chr.2L: 15854126 - 15856450 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028902
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273547.1 NP_001260476.1 2 - 3 875 76
NM_078853.3 NP_523577.1 2 - 3 733 76
GenBank mRNA AY058398.1 - 2 - 3 720 76

View Full Product Details