Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dhc36C
Entrez Gene ID
35061
Gene Name
Dynein heavy chain at 36C
Gene Aliases
CG5526, DHC, DHC7A, Dhc, Dhc36c, Dmel\CG5526, Dmel_CG5526, dhc
Chromosome Location
Chr.2L: 17501401 - 17525515 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013810
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382179.1 NP_001369131.1 15 - 16 8608 119
NM_078867.4 NP_523591.2 15 - 16 8608 119

View Full Product Details