Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tou
Entrez Gene ID
36241
Gene Name
toutatis
Gene Aliases
CG10897, Dm Tou, Dmel\CG10897, Dmel_CG10897, EP(2)0622, EP0622, EP622, TIP5, TOU, Tou, VI, anon-48Ad, gene VI
Chromosome Location
Chr.2R: 11577485 - 11616045 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033636
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103800.3 NP_001097270.1 3 - 4 899 66
NM_001273969.1 NP_001260898.1 3 - 4 600 66
NM_001273970.1 NP_001260899.1 4 - 5 865 66
NM_078977.5 NP_523701.3 3 - 4 899 66

View Full Product Details