Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Vps25
- Entrez Gene ID
-
35847
- Gene Name
- Vacuolar protein sorting 25
- Gene Aliases
-
CG14750, Dmel\CG14750, Dmel_CG14750, ESCRT-I, ESCRT-II, VPS25, dVps25, l(2)44Db, l(2)k08904, vps25
- Chromosome Location
-
Chr.2R: 8625451 - 8626180 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0022027
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136554.3 |
NP_610398.1 |
1 - 2 |
121 |
68 |
GenBank mRNA |
AY089657.1 |
- |
1 - 2 |
120 |
68 |
View Full Product Details