Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pds5
Entrez Gene ID
36286
Gene Name
precocious dissociation of sisters 5
Gene Aliases
CG17509, Dmel\CG17509, Dmel_CG17509, PDS5, Pds5, anon-WO0118547.183, dPds5, l(2)k13312
Chromosome Location
Chr.2R: 12027985 - 12033725 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260012
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259342.2 NP_001246271.1 4 - 5 1204 74
NM_136875.4 NP_610719.1 4 - 5 946 74
GenBank mRNA BT053744.1 - 4 - 5 926 74

View Full Product Details