Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gol
Entrez Gene ID
38006
Gene Name
goliath
Gene Aliases
CG2679, Dmel\CG2679, Dmel_CG2679, Gol, anon-G1, dgol, g1, meso
Chromosome Location
Chr.2R: 25072627 - 25084347 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004919
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169829.3 NP_001163300.1 5 - 6 1529 65
NM_001259588.2 NP_001246517.1 5 - 6 1529 65
NM_079140.5 NP_523864.3 5 - 6 1529 65
NM_166705.3 NP_726508.1 5 - 6 1529 65
GenBank mRNA AY069169.1 - 5 - 6 1502 65

View Full Product Details