The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001274267.1 | NP_001261196.1 | 1 - 2 | 271 | 68 |
NM_079146.3 | NP_523870.2 | 1 - 2 | 271 | 68 | |
GenBank mRNA | AY119639.1 | - | 1 - 2 | 268 | 68 |
BT029976.1 | - | 1 - 2 | 259 | 68 | |
BT120114.1 | - | 1 - 2 | 259 | 68 |