Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acp63F
Entrez Gene ID
38448
Gene Name
Accessory gland protein 63F
Gene Aliases
Acp63, Acp63F/64A, Acp63f, CG10852, Dmel\CG10852, Dmel_CG10852, acp63F
Chromosome Location
Chr.3L: 3815839 - 3816354 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015585
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316457.1 NP_001303386.1 2 - 3 189 71
NM_079182.3 NP_523906.2 2 - 3 189 71
GenBank mRNA BT053707.1 - 2 - 3 181 71

View Full Product Details