Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tak1
- Entrez Gene ID
-
39659
- Gene Name
- TGF-beta activated kinase 1
- Gene Aliases
-
CG1388, CG18492, D-TAK, D-tak, D10, DTAK1, DTak, DTak1, DmTak1, Dmel\CG18492, Dmel_CG18492, TAK, TAK-1, TAK1, Tak, Tak 1, dTAK, dTAK-1, dTAK1, dTak, dTak1, dtak1, tak, tak1
- Chromosome Location
-
Chr.X: 20515908 - 20524928 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026323
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079356.3 |
NP_524080.1 |
1 - 2 |
1032 |
71 |
GenBank mRNA |
AY051953.1 |
- |
1 - 2 |
1005 |
71 |
View Full Product Details