Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smn
Entrez Gene ID
39844
Gene Name
survival motor neuron
Gene Aliases
CG16725, DmSMN, Dmel\CG16725, Dmel_CG16725, SMN, dSMN, dSmn, l(3)73Ao, smn
Chromosome Location
Chr.3L: 16580398 - 16581547 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036641
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275025.1 NP_001261954.1 2 - 2 977 77
NM_079388.4 NP_524112.1 1 - 1 770 77
GenBank mRNA AF296281.1 - 1 - 1 735 77
AY058529.1 - 1 - 1 734 77

View Full Product Details