Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fng
Entrez Gene ID
40314
Gene Name
fringe
Gene Aliases
CG10580, D-Fng, D-fng, Dfng, Dmel\CG10580, Dmel_CG10580, Fng, Frg, fg, frg, l(3)rG554
Chromosome Location
Chr.3L: 20925891 - 20949512 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011591
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079467.3 NP_524191.1 5 - 6 1038 72
GenBank mRNA AY070927.1 - 5 - 6 1035 72
L35770.1 - 5 - 6 938 72

View Full Product Details