Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stj
Entrez Gene ID
36526
Gene Name
straightjacket
Gene Aliases
CG12295, Dmel\CG12295, Dmel_CG12295, alpha[[2]]delta, alpha[[2]]delta-3, dalpha[[2]]delta-3, l(2)k10814
Chromosome Location
Chr.2R: 13805794 - 13819571 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261041
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259374.2 NP_001246303.1 22 - 23 3595 56
NM_001259375.2 NP_001246304.1 20 - 21 3884 56
NM_137058.4 NP_610902.1 19 - 20 3827 56
GenBank mRNA AY069830.1 - 19 - 20 4147 56

View Full Product Details