Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fas2
Entrez Gene ID
31364
Gene Name
Fasciclin 2
Gene Aliases
1D4, Ab 1D4, CG3665, CT12301, Dmel\CG3665, Dmel_CG3665, EG:EG0007.3, FAS 11, FAS II, FAS2, FASII, Fas, Fas 2, Fas II, Fas-II, FasII, Fasc II, FascII, Fasii, MAB1D4, anon-EST:Liang-1.60, clone 1.60, fas II, fas-II, fas2, fasII, fasciclin II, l(1)G0032, l(1)G0048, l(1)G0081, l(1)G0293, l(1)G0336, mAB1D4, mAb 1D4, mAb1D4
Chromosome Location
Chr.X: 4132887 - 4206093 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000635
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272306.1 NP_001259235.1 4 - 5 826 106
NM_001272308.2 NP_001259237.1 5 - 6 871 106
NM_001297925.1 NP_001284854.1 4 - 5 826 106
NM_001297926.1 NP_001284855.1 4 - 5 826 106
NM_080327.3 NP_525066.1 4 - 5 826 106
NM_167004.2 NP_726911.1 4 - 5 826 106
NM_167005.2 NP_726912.1 4 - 5 826 106
GenBank mRNA BT014661.1 - 4 - 5 809 106
M77165.1 - 4 - 5 767 106
M77166.1 - 4 - 5 767 106

View Full Product Details