Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wit
Entrez Gene ID
44096
Gene Name
wishful thinking
Gene Aliases
1262/15, ALK3/BMPRII, BMP, CG10776, Dmel\CG10776, Dmel_CG10776, SE20, STK-D, Stk-D, WIT, Wit, Wit-C, l(3)64Aa, l(3)S126215, l(3)SH12, wiw
Chromosome Location
Chr.3L: 4064152 - 4071717 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024179
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274466.1 NP_001261395.1 5 - 6 2080 80
NM_079953.4 NP_524692.3 5 - 6 2080 80
GenBank mRNA AY119514.1 - 5 - 6 1865 80

View Full Product Details