Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- clu
- Entrez Gene ID
-
36793
- Gene Name
- clueless
- Gene Aliases
-
CG8443, Clu, Dmel\CG8443, Dmel_CG8443, anon-WO0118547.143
- Chromosome Location
-
Chr.2R: 16186584 - 16194277 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034087
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137251.3 |
NP_611095.1 |
1 - 2 |
555 |
93 |
View Full Product Details