Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ste24a
- Entrez Gene ID
-
36908
- Gene Name
- prenyl protease type I
- Gene Aliases
-
CG9000, Dmel\CG9000, Dmel\Ste24, Dmel\Ste24a, Dmel_CG9000, Ste24a
- Chromosome Location
-
Chr.2R: 17000244 - 17001941 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034176
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137331.3 |
NP_611175.1 |
1 - 2 |
175 |
73 |
GenBank mRNA |
AY069332.1 |
- |
1 - 2 |
163 |
73 |
View Full Product Details