Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIP1
Entrez Gene ID
43981
Gene Name
DISCO Interacting Protein 1
Gene Aliases
CG17686, Dmel\CG17686, Dmel_CG17686, TO34, TO67, UbxBP1, clot 10495, clot 2020, dip1, dip1a, klt
Chromosome Location
Chr.X: 21624796 - 21630148 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024807
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_167772.2 NP_728449.1 1 - 2 170 63
GenBank mRNA AF175711.2 - 1 - 2 153 63
BT099548.1 - 1 - 2 148 63

View Full Product Details