Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nord
Entrez Gene ID
37882
Gene Name
CG30418-PA
Gene Aliases
CG13574, CG30418, CG3372, Dmel\CG30418, Dmel_CG30418, E3272
Chromosome Location
Chr.2R: 24243255 - 24255094 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0050418
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138056.3 NP_611900.1 7 - 8 1990 100
GenBank mRNA BT133485.1 - 7 - 8 1982 100

View Full Product Details