Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4d14
Entrez Gene ID
45706
Gene Name
CG3540-PA
Gene Aliases
4d14, CG3540, CYP4D14, Cyp4D14, Dmel\CG3540, Dmel_CG3540, EG:152A3.2
Chromosome Location
Chr.X: 2173415 - 2175315 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023541
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272250.2 NP_001259179.1 2 - 3 708 60
NM_143763.4 NP_652020.1 2 - 3 699 60
GenBank mRNA BT050587.1 - 2 - 3 697 60

View Full Product Details