Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
obst-I
Entrez Gene ID
317966
Gene Name
obstructor-I
Gene Aliases
CG13805, CG32304, Dmel\CG32304, Dmel_CG32304
Chromosome Location
Chr.3L: 2267858 - 2268590 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052304
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_167943.3 NP_728731.1 1 - 1 608 76
GenBank mRNA BT100064.5 - 1 - 1 596 76

View Full Product Details