Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr64a
- Entrez Gene ID
-
117481
- Gene Name
- Gustatory receptor 64a
- Gene Aliases
-
CG14986, CG32261, Dmel64a, Dmel\CG32261, Dmel_CG32261, GR64a, GRLU.3, Gr64, Gr64A1, GrLU3, LU.3
- Chromosome Location
-
Chr.3L: 4026769 - 4028600 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0045479
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168048.2 |
NP_728920.1 |
2 - 3 |
390 |
80 |
View Full Product Details