Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DnaJ-1
Entrez Gene ID
38643
Gene Name
DnaJ-like-1
Gene Aliases
CG10578, DNAJ-1, DNAJ1, DROJ1, DmHsp40, Dmel\CG10578, Dmel_CG10578, DnaJ1, DnaJ1 64EF, Dnaj-1, DroJ1, EU3500, HSP40, HSP40/HDJ1, Hdj1, Hsp-40, Hsp40, anon-WO0140519.166, anon-WO0172774.135, dHDJ1, dHDJ1/HSP40, dHSP40, dHdj1, dhdJ1, dhdj-1, dhdj1, dnaJ-1, dnaj-1, droj1, hsp40
Chromosome Location
Chr.3L: 5750029 - 5752189 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263106
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079212.4 NP_523936.2 2 - 2 747 50
NM_168128.3 NP_729086.1 1 - 1 992 50
GenBank mRNA AY058788.1 - 2 - 2 666 50

View Full Product Details