Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4d20
Entrez Gene ID
38311
Gene Name
CG16761-PA
Gene Aliases
4d20, CG16761, CYP4d20, Dmel\CG16761, Dmel_CG16761, RH59895
Chromosome Location
Chr.3L: 2476259 - 2478448 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035344
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139466.5 NP_647723.2 2 - 3 691 59
GenBank mRNA BT133316.1 - 2 - 3 687 59

View Full Product Details