Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fhos
Entrez Gene ID
39004
Gene Name
Formin homology 2 domain containing
Gene Aliases
CG32025, CG32030, CG42610, CG5775, CG5797, Dmel\CG42610, Dmel_CG42610, dFHOS, desert, jaz, knit, l(3)01629
Chromosome Location
Chr.3L: 8753619 - 8798719 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266084
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202142.2 NP_001189071.1 2 - 3 508 62
NM_001202143.2 NP_001189072.1 2 - 3 508 62
NM_001259754.2 NP_001246683.1 2 - 3 508 62
NM_168293.3 NP_729410.1 2 - 3 508 62
NM_168294.3 NP_729411.1 2 - 3 508 62
GenBank mRNA BT050456.1 - 2 - 3 474 62

View Full Product Details