Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nxf3
Entrez Gene ID
317878
Gene Name
Nuclear export factor 3
Gene Aliases
CG32135, Dmel\CG32135, Dmel_CG32135, NXF3, nxf3
Chromosome Location
Chr.3L: 13965203 - 13973067 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263232
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300135.1 NP_001287064.2 2 - 2 3104 114
NM_168566.2 NP_729938.2 2 - 2 3104 114
GenBank mRNA BT021396.1 - 2 - 2 3078 114

View Full Product Details