Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Smyd3
- Entrez Gene ID
-
44554
- Gene Name
- SET and MYND domain containing, class 3
- Gene Aliases
-
Bzd, CG13761, Dmel\CG13761, Dmel_CG13761, EG:BACR7C10.4
- Chromosome Location
-
Chr.X: 2606680 - 2608876 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011566
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080029.4 |
NP_524768.2 |
4 - 5 |
1239 |
60 |
GenBank mRNA |
BT060437.1 |
- |
4 - 5 |
1239 |
60 |
View Full Product Details