The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272656.2 | NP_001259585.1 | 4 - 5 | 2895 | 78 |
NM_001272657.2 | NP_001259586.1 | 4 - 5 | 2815 | 78 | |
NM_001298376.1 | NP_001285305.1 | 4 - 5 | 2917 | 78 | |
NM_132858.3 | NP_573086.2 | 4 - 5 | 2920 | 78 | |
GenBank mRNA | AY118524.1 | - | 4 - 5 | 2895 | 78 |