Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rrp45
Entrez Gene ID
32665
Gene Name
CG9606-PA
Gene Aliases
CG9606, Dmel\CG9606, Dmel_CG9606, dRrp45, rrp45
Chromosome Location
Chr.X: 16789405 - 16791266 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030789
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298424.1 NP_001285353.1 1 - 2 590 86
NM_132935.3 NP_573163.1 1 - 2 590 86
GenBank mRNA AY089277.1 - 1 - 2 479 86

View Full Product Details