Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chd64
Entrez Gene ID
38490
Gene Name
CG14996-PB
Gene Aliases
CG-14996, CG14996, CT34849, Dmel\CG14996, Dmel_CG14996, anon-EST:Liang-1.80, clone 1.80
Chromosome Location
Chr.3L: 4114289 - 4122388 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035499
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139603.3 NP_647860.1 1 - 2 230 64
GenBank mRNA AF217286.1 - 1 - 2 203 64
AY060355.1 - 1 - 2 206 64

View Full Product Details