Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LanA
- Entrez Gene ID
-
38723
- Gene Name
- Laminin A
- Gene Aliases
-
5]], CG10236, CT28769, Dmel\CG10236, Dmel_CG10236, FBgn0002526, FBtr0077014, LM-A/alpha1, LMalpha1, Lam, Lam-A, LamA, Lan, alpha3/5, alpha[[3, hdln, lamA, laminin alpha3/5, lanA
- Chromosome Location
-
Chr.3L: 6203831 - 6218022 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002526
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057269.3 |
NP_476617.1 |
13 - 14 |
9677 |
68 |
View Full Product Details