Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CalpA
Entrez Gene ID
37232
Gene Name
Calpain-A
Gene Aliases
CG18152, CG7563, Dmel\CG7563, Dmel_CG7563, EST C, calpA
Chromosome Location
Chr.2R: 19425113 - 19430564 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0012051
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103908.2 NP_001097378.1 9 - 10 1678 67
NM_001299684.1 NP_001286613.1 10 - 11 1723 67
NM_057699.5 NP_477047.1 9 - 10 1586 67
NM_057700.4 NP_477048.1 9 - 10 1678 67
GenBank mRNA AY051678.1 - 8 - 9 1562 67

View Full Product Details