Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dare
Entrez Gene ID
36203
Gene Name
defective in the avoidance of repellents
Gene Aliases
AR, CG12390, Dare, Dmel\CG12390, Dmel_CG12390, W8
Chromosome Location
Chr.2R: 11290994 - 11293174 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015582
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057802.6 NP_477150.1 3 - 4 916 56
GenBank mRNA AF168685.1 - 3 - 4 892 56
BT083432.1 - 3 - 4 913 56

View Full Product Details