Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4p1
Entrez Gene ID
45524
Gene Name
Cytochrome P450-4p1
Gene Aliases
4p1, CG10842, CYP4p1, Cyp4P1, Dmel\CG10842, Dmel_CG10842, P-450, P450, anon-WO0140519.35
Chromosome Location
Chr.2R: 9240041 - 9242139 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015037
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080089.3 NP_524828.1 1 - 2 246 64
GenBank mRNA AY071584.1 - 1 - 2 244 64

View Full Product Details