Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PpD6
- Entrez Gene ID
-
49780
- Gene Name
- Protein phosphatase D6
- Gene Aliases
-
CG8822, D6, Dmel\CG8822, Dmel_CG8822, PP1
- Chromosome Location
-
Chr.2L: 3109672 - 3110998 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0005779
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080208.3 |
NP_524947.1 |
1 - 1 |
1002 |
77 |
View Full Product Details