Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- toc
- Entrez Gene ID
-
33526
- Gene Name
- toucan
- Gene Aliases
-
BA34, BcDNA:LD27161, CG9660, Dmel\CG9660, Dmel_CG9660, Toc, l(2)01361, l(2)05527, l(2)K08224, l(2)k08224
- Chromosome Location
-
Chr.2L: 3068345 - 3144786 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015600
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_164525.2 |
NP_722876.1 |
1 - 2 |
231 |
115 |
GenBank mRNA |
BT099598.1 |
- |
1 - 2 |
231 |
115 |
View Full Product Details