Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ance-2
- Entrez Gene ID
-
34806
- Gene Name
- Ance-2-PA
- Gene Aliases
-
CG16869, CG31844, Dmel\CG16869, Dmel_CG16869, ance-2
- Chromosome Location
-
Chr.2L: 13909333 - 13911719 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032535
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176028.2 |
NP_788042.1 |
2 - 3 |
1272 |
69 |
View Full Product Details