Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- spn-E
- Entrez Gene ID
-
41919
- Gene Name
- spindle E
- Gene Aliases
-
CG3158, Dmel\CG3158, Dmel_CG3158, SPN-E, Spn-E, SpnE, fs(1)hls, fs(3)hls, hes, hls, homeless, spin-E, spn E, spn-E/hls, spnE
- Chromosome Location
-
Chr.3R: 15837419 - 15844415 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003483
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057393.4 |
NP_476741.1 |
5 - 6 |
2019 |
97 |
GenBank mRNA |
BT100306.1 |
- |
5 - 6 |
1955 |
97 |
View Full Product Details