Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tok
Entrez Gene ID
42944
Gene Name
tolkin
Gene Aliases
CG6863, CT21251, Dmel\CG6863, Dmel_CG6863, TLR, TOK, Tlr, Tlr-1, Tok, tldr-1, tldr1, tlr, tlr-1, tlr1
Chromosome Location
Chr.3R: 24719697 - 24749125 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004885
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300581.1 NP_001287510.1 11 - 12 3944 102
NM_057531.4 NP_476879.1 11 - 12 3948 102
NM_170168.2 NP_733035.1 11 - 12 3827 102
GenBank mRNA AY051585.1 - 11 - 12 3934 102

View Full Product Details