Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tmod
Entrez Gene ID
43633
Gene Name
tropomodulin
Gene Aliases
1309/10, CG11493, CG1539, CG15540, CT3919, CT41421, Dmel\CG1539, Dmel_CG1539, E42, Tmod, l(3)00848, l(3)S130910, spdo
Chromosome Location
Chr.3R: 30532607 - 30578905 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0082582
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104512.4 NP_001097982.1 9 - 10 2408 83
NM_001170304.3 NP_001163775.1 6 - 7 1933 83
NM_001170305.3 NP_001163776.1 7 - 8 2216 83
NM_001170306.2 NP_001163777.1 7 - 8 1969 83
NM_001202390.1 NP_001189319.1 8 - 9 2448 83
NM_001202391.2 NP_001189320.1 7 - 8 1878 83
NM_001260444.2 NP_001247373.1 9 - 10 2700 83
NM_001276183.1 NP_001263112.1 7 - 8 2398 83
NM_057683.5 NP_477031.1 6 - 7 1753 83
NM_057684.4 NP_477032.1 6 - 7 1724 83
NM_170503.4 NP_733382.1 6 - 7 2122 83
NM_170504.4 NP_733383.1 7 - 8 1901 83
NM_170505.4 NP_733384.1 7 - 8 1986 83
NM_170506.3 NP_733385.1 7 - 8 1932 83
GenBank mRNA AY094727.1 - 6 - 7 1724 83
BT099728.1 - 8 - 9 2449 83
BT133205.1 - 6 - 7 1725 83
BT133473.1 - 9 - 10 2400 83
U92490.1 - 6 - 7 1762 83

View Full Product Details