Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt4
Entrez Gene ID
40876
Gene Name
Synaptotagmin 4
Gene Aliases
CG10047, D.Syt IV, Dmel\CG10047, Dmel_CG10047, SYT4, Syt IV, SytIV, dsyt4, syt 4, syt4, sytIV
Chromosome Location
Chr.3R: 7266065 - 7270809 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028400
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058116.5 NP_477464.1 2 - 3 231 59
GenBank mRNA AF181098.1 - 2 - 3 193 59
BT133170.1 - 2 - 3 235 59

View Full Product Details