Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or85b
- Entrez Gene ID
-
41007
- Gene Name
- Odorant receptor 85b
- Gene Aliases
-
85A.1, 85b, CG11735, DOR115, DmOR85b, DmOr85b, Dmel\CG11735, Dmel_CG11735
- Chromosome Location
-
Chr.3R: 8510077 - 8511358 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037590
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079555.2 |
NP_524279.2 |
2 - 3 |
1116 |
139 |
View Full Product Details